missing translation for 'onlineSavingsMsg'
Learn More

XL388, TRC

Product Code. 30453787
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
Product Code. Quantity unitSize
30453787 10 mg 10mg
30449116 2.5 mg 2.5mg
2 options
This item is not returnable. View return policy

Product Code. 30453787

Brand: Toronto Research Chemicals TRCX74622010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1251156-08-7
Formula Weight 455.1315
InChI Formula InChI=1S/C23H22FN3O4S/c1-14-18(5-7-20(22(14)24)32(2,29)30)23(28)27-9-10-31-19-6-3-15(11-17(19)13-27)16-4-8-21(25)26-12-16/h3-8,11-12H,9-10,13H2,1-2H3,(H2,25,26)
IUPAC Name [7-(6-aminopyridin-3-yl)-3,5-dihydro-2H-1,4-benzoxazepin-4-yl]-(3-fluoro-2-methyl-4-methylsulfonylphenyl)methanone
Molecular Formula C23 H22 F N3 O4 S
Molecular Weight (g/mol) 455.5
SMILES Cc1c(F)c(ccc1C(=O)N2CCOc3ccc(cc3C2)c4ccc(N)nc4)S(=O)(=O)C
Synonym (7-(6-Aminopyridin-3-yl)-2,3-dihydrobenzo[f][1,4]oxazepin-4(5H)-yl)(3-fluoro-2-methyl-4-(methylsulfonyl)phenyl)methanone, [7-(6-Amino-3-pyridinyl)-2,3-dihydro-1,4-benzoxazepin-4(5H)-yl][3-fluoro-2-methyl-4-(methylsulfonyl)phenyl]-methanone
Chemical Name or Material XL388

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.