Learn More
Tocris Bioscience™ XL 388
Potent and selective mTOR inhibitor; antitumor
Specifications
Purity >98%
View More Specs
Description
Description
Potent and selective mTOR inhibitor (IC50 = 9.9 nM). Inhibits mTOR activity in an ATP-competitive manner. Exhibits >300-fold selectivity for mTOR over PI 3-K and a range of other kinases. Displays antitumor activity in athymic nude mice implanted with tumor xenografts.
Specifications
Specifications
>98%