missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XKR8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572820
This item is not returnable.
View return policy
Description
XKR8 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Promotes phosphatidylserine exposure on apoptotic cell surface, possibly by mediating phospholipid scrambling. Phosphatidylserine is a specific marker only present at the surface of apoptotic cells and acts as a specific signal for engulfment. Has no effect on calcium-induced exposure of phosphatidylserine. Activated upon caspase cleavage, suggesting that it does not act prior the onset of apoptosis.
Specifications
| XKR8 | |
| Polyclonal | |
| Unconjugated | |
| XKR8 | |
| hXkr8; RP11-460I13.3; X Kell blood group precursor-related family, member 8; XK related 8; XK, Kell blood group complex subunit-related family, member 8; XKR8; XK-related protein 8; XK-related protein 8, processed form; X-linked Kx blood group related 8; XRG8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9H6D3 | |
| XKR8 | |
| XKR8 antibody for a 15 amino acid peptide near the carboxy terminus of human XKR8. The immunogen is within the last 50 amino acids of XKR8. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur