missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XK X-linked Kx blood group Antibody, mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
XK X-linked Kx blood group Polyclonal antibody specifically detects XK X-linked Kx blood group in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Dilution | Western Blot, Immunocytochemistry/ Immunofluorescence |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?