missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XK X-linked Kx blood group Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-05963CL1
This item is not returnable.
View return policy
Description
XK X-linked Kx blood group Polyclonal antibody specifically detects XK X-linked Kx blood group in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| XK X-linked Kx blood group | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Kell blood group precursor (McLeod phenotype), Kell complex 37 kDa component, KX, Kx antigen, membrane transport protein XK, X1k, XK, Kell blood group complex subunit (McLeod syndrome), XKR1XK-related protein 1, X-linked Kx blood group (McLeod syndrome), XRG1 | |
| Produced in rabbits immunized with E. coli-derived Human XK X-linked Kx blood group fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 7504 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction