missing translation for 'onlineSavingsMsg'
Learn More

XK X-linked Kx blood group Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18353296 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353296 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353296 Supplier Novus Biologicals Supplier No. NBP305963AF350

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XK X-linked Kx blood group Polyclonal specifically detects XK X-linked Kx blood group in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XK X-linked Kx blood group
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Kell blood group precursor (McLeod phenotype), Kell complex 37 kDa component, KX, Kx antigen, membrane transport protein XK, X1k, XK, Kell blood group complex subunit (McLeod syndrome), XKR1XK-related protein 1, X-linked Kx blood group (McLeod syndrome), XRG1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human XK X-linked Kx blood group fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7504
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.