missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XIAP Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10037-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.Specifications
| XIAP | |
| Polyclonal | |
| Unconjugated | |
| XIAP | |
| API3, BIRC4, hIAP 3, hIAP3, hILP, IAP 3, IAP like protein, IAP3, ILP, ILP1, MIHA, X linked IAP, XIAP, XLP2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 331, 63879 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P98170, Q9R0I6 | |
| XIAP | |
| XIAP Fusion Protein Ag0022 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction