missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XIAP Polyclonal Antibody

Rabbit Polyclonal Antibody
£155.00 - £430.00
Specifications
| Antigen | XIAP |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17730914
![]() |
Invitrogen™
PA5119509 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17773234
|
Invitrogen™
PA5121839 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XIAP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP+ phosphate to ATP. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits. While the F0 consists of a, b, and c subunits.Specifications
| XIAP | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Pig | |
| XIAP | |
| 1110015C02Rik; Aipa; Api3; apoptosis inhibitor protein 3; baculoviral IAP repeat-containing 4; baculoviral IAP repeat-containing protein 4; BIR4; Birc4; E3 ubiquitin-protein ligase XIAP; hIAP3; hIAP-3; HILP; IAP homolog A; IAP3; IAP-3; IAP-like protein; ILP; ILP1; ILP-1; Inhibitor of apoptosis protein 3; MIHA; riap3; RIAP-3; RING-type E3 ubiquitin transferase XIAP; RP1-315G1.5; XIAP; X-linked IAP; X-linked inhibitor of apoptosis; X-linked inhibitor of apoptosis protein; X-linked inhibitor of apoptosis, E3 ubiquitin protein ligase; XLP2 | |
| XIAP | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P98170 | |
| 100037300, 331 | |
| Recombinant protein X-linked Inhibitor Of Apoptosis Protein. The antigen corresponds to amino acid range 1-497 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title