missing translation for 'onlineSavingsMsg'
Learn More

Xenon difluoride, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11334707
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorinating agent

Used as Fluorinating agent and oxidizing agent, gaseous etchant for silicon, particularly in the production of microelectromechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-36-9
Molecular Formula F2Xe
Molecular Weight (g/mol) 169.29
MDL Number MFCD00040538
InChI Key IGELFKKMDLGCJO-UHFFFAOYSA-N
Synonym xenon difluoride, xenon fluoride, xenon fluoride xef2, unii-6poj14981p, bis fluoranyl xenon, xef2, acmc-20aj9j, xenon difluoride trace metals basis
PubChem CID 83674
IUPAC Name difluoroxenon
SMILES F[Xe]F

Specifications

Melting Point 127°C to 129°C
Physical Form Crystalline Granules
Quantity 10 g
Assay Percent Range 99.5%, metal basis
Solubility Information Decomposes in water.
Formula Weight 169.3
Packaging PTFE
Sensitivity Moisture sensitive
Density 4.32
Chemical Name or Material Xenon Difluoride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'