missing translation for 'onlineSavingsMsg'
Learn More

XEN907, MedChemExpress

Product Code. 16460448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16460448 Supplier Medchem Express Supplier No. HY19958
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress XEN907 is a potent and spirooxindole blocker of NaV1.7, with an IC50 of 3 nM. XEN907 also inhibits CYP3A4 in a recombinant human enzyme assay. XEN907 can be used for the research of pain.

XEN907 is a potent and spirooxindole blocker of NaV1.7, with an IC50 of 3 nM. XEN907 also inhibits CYP3A4 in a recombinant human enzyme assay. XEN907 can be used for the research of pain.

Specifications

Chemical Name or Material XEN907
CAS 912656-34-9
Color White
Purity Grade Notes Research
Molecular Formula C21H21NO4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (284.58 mM; Need ultrasonic)
SMILES O=C(C12COC3=C1C=C(OCO4)C4=C3)N(CCCCC)C5=C2C=CC=C5
Molecular Weight (g/mol) 351.4
Formula Weight 351.4
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.