missing translation for 'onlineSavingsMsg'
Learn More

XEN723, MedChemExpress

Product Code. p-200097382
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16490559

Brand: Medchem Express HY100249

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress XEN723 is a novel and potent thiazolylimidazolidinone inhibitor of Stearoyl-CoA Desaturase (SCD1) with IC50s of 45 and 524 nM in mouse and HepG2 cell, respectively.

XEN723 is a novel and potent thiazolylimidazolidinone inhibitor of Stearoyl-CoA Desaturase (SCD1) with IC50s of 45 and 524 nM in mouse and HepG2 cell, respectively.

Specifications

Chemical Name or Material XEN723
CAS 1072803-08-7
Color White
Purity Grade Notes Research
Molecular Formula C21H20FN5O2S
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (235.03 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N=C(N2CCN(CC3=CC=C(F)C=C3)C2=O)S1)NCC4=CC=CN=C4
Molecular Weight (g/mol) 425.48
Formula Weight 425.48
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.