missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XE7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32138
This item is not returnable.
View return policy
Description
XE7 Polyclonal specifically detects XE7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| XE7 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| AKAP17A | |
| Recombinant protein encompassing a sequence within the C-terminus region of human XE7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| A kinase (PRKA) anchor protein 17A, AKAP-17A, A-kinase anchor protein 17A, B-lymphocyte antigen, B-lymphocyte surface antigen, CCDC133, CXYorf3, DXYS155EPRKA17A, MGC39904, Protein kinase A-anchoring protein 17A, Protein XE7, pseudoautosomal gene XE7, SFRS17AMGC125366, Splicing factor, arginine/serine-rich 17AFLJ98315,721PMGC125365, XE7chromosome X and Y open reading frame 3, XE7Y | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8227 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction