missing translation for 'onlineSavingsMsg'
Learn More

XE 991 Dihydrochloride, TRC

Product Code. 30451502
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451502 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451502 Supplier Toronto Research Chemicals Supplier No. TRCX750935100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material XE 991 Dihydrochloride
CAS 122955-13-9
Molecular Formula C26H20N2O • 2(HCl)
Quantity 100 mg
Synonym DR 960418, XE 991, 10,10-Bis(4-pyridinylmethyl)-9(10H)-anthracenone
InChI Formula InChI=1S/C26H20N2O.2ClH/c29-25-21-5-1-3-7-23(21)26(17-19-9-13-27-14-10-19,18-20-11-15-28-16-12-20)24-8-4-2-6-22(24)25;;/h1-16H,17-18H2;2*1H
SMILES O=C1C2=C(C(CC3=CC=NC=C3)(C4=CC=CC=C41)CC5=CC=NC=C5)C=CC=C2.Cl.Cl
IUPAC Name 10,10-bis(pyridin-4-ylmethyl)anthracen-9(10H)-one dihydrochloride
Molecular Weight (g/mol) 449.37
Formula Weight 448.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.