missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55156-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to XDH.
XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.Specifications
| XDH | |
| Polyclonal | |
| Unconjugated | |
| XDH | |
| xanthine dehydrogenase, Xanthine Oxidase, Xanthine oxidoreductase, XD, XDH, XDHA, XO, XOR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22436, 497811, 7498 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P22985, P47989, Q00519 | |
| Xdh | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction