missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XDH Monoclonal Antibody (1C8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA534898
This item is not returnable.
View return policy
Description
Positive Control: Human liver tissue, human kidney tissue, LOVO.
XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.
Specifications
| XDH | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P47989 | |
| XDH | |
| Synthetic peptide within Human xanthine dehydrogenase aa 1,082-1,131. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 1C8 | |
| Unconjugated | |
| XDH | |
| Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 7498 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction