missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ XCT 790

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective ERRα inverse agonist (IC50 = ∽400 nM). Displays no antagonist activity at ERRγ or ERα at concentrations below 10 μM. Interferes with PPARγ coactivator-1α (PGC-1α)/ERRα-dependent signaling: inhibits PGC-1α induction of ERRα and MAO-B gene expression and downregulates the constitutive transcriptional activity of ERRα in numerous cell lines.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 596.42 |
| Purity | >99% |
| Molecular Weight (g/mol) | 596.428 |
| Chemical Name or Material | XCT 790 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction