missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XCR1/CCXCR1 Antibody (1097A) [PerCP], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8571C
This item is not returnable.
View return policy
Description
XCR1/CCXCR1 Monoclonal antibody specifically detects XCR1/CCXCR1 in Human samples. It is validated for Flow Cytometry
Specifications
| XCR1/CCXCR1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1097A | |
| Flow Cytometry | |
| CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor | |
| E. coli-derived recombinant human XCR1, Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267, Accession # P46094 | |
| 0.1 mL | |
| Chemokines and Cytokines, GPCR, Immunology, Innate Immunity | |
| 2829 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction