missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XCL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | XCL2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606897
|
Novus Biologicals
NBP3-06178-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605048
|
Novus Biologicals
NBP3-06178-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XCL2 Polyclonal antibody specifically detects XCL2 in Human samples. It is validated for ELISASpecifications
| XCL2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C motif chemokine 2, chemokine (C motif) ligand 2, cytokine SCM-1 beta, SCM-1b, SCM1B, SCYC2XC chemokine ligand 2, small inducible cytokine subfamily C, member 2 | |
| Produced in rabbits immunized with purified, recombinant Human XCL2 (Uniprot#: Q9UBD3; Met1-Gly114) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 6846 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title