missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XCL2 Antibody (06), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06177CL1
This item is not returnable.
View return policy
Description
XCL2 Monoclonal antibody specifically detects XCL2 in Human samples. It is validated for ELISA
Specifications
| XCL2 | |
| Monoclonal | |
| CoraFluor 1 | |
| C motif chemokine 2, chemokine (C motif) ligand 2, cytokine SCM-1 beta, SCM-1b, SCM1B, SCYC2XC chemokine ligand 2, small inducible cytokine subfamily C, member 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human XCL2 (Uniprot#: Q9UBD3; Met1-Gly114). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 06 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6846 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction