missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XCL1/Lymphotactin Rabbit anti-Mouse, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90673-100UL
This item is not returnable.
View return policy
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Mouse samples. It is validated for ELISA, Sandwich ELISA Capture.
Spezifikation
| XCL1/Lymphotactin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 2 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 | |
| ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| Recombinant Mouse XCL1/Lymphotactin protein | |
| 100 μg | |
| Biologically Active Proteins, Immunology, Innate Immunity | |
| 6375 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur