missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XCL1/Lymphotactin Antibody (MM0604-4G24), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12004MFV610
This item is not returnable.
View return policy
Description
XCL1/Lymphotactin Monoclonal antibody specifically detects XCL1/Lymphotactin in Human samples. It is validated for Western Blot
Specifications
| XCL1/Lymphotactin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0604-4G24 | |
| Western Blot | |
| ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| Human recombinant XCL1/Lymphotactin | |
| 0.1 mL | |
| Biologically Active Proteins, Immunology, Innate Immunity | |
| 6375 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction