missing translation for 'onlineSavingsMsg'
Learn More

XCL1/Lymphotactin Antibody (002), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18361801 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361801 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361801 Supplier Novus Biologicals Supplier No. NBP290673AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XCL1/Lymphotactin
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse XCL1/Lymphotactin (P47993; Met1-Gly114).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 6375
Test Specificity No cross-reactivity in ELISA with: Human XCL1 Human CXCL1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.