missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XCL1/Lymphotactin Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90947CL1
This item is not returnable.
View return policy
Description
XCL1/Lymphotactin Monoclonal antibody specifically detects XCL1/Lymphotactin in Cynomolgus Monkey samples. It is validated for ELISA
Specifications
| XCL1/Lymphotactin | |
| Monoclonal | |
| CoraFluor 1 | |
| ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus XCL1/Lymphotactin (Accession#: ; Met1-Gly114). | |
| 0.1 mL | |
| Biologically Active Proteins, Immunology, Innate Immunity | |
| 6375 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction