missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge BEH130 C18 Column

The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations.

Brand:  Waters Corporation 186003561

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13175179

  • £1005.00 / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • The small-pore (130 Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Specifications

Specifications

1 mm
6000 psi (415 Bar)
3.5 μm
1/Ea.
Yes
Peptides
100 mm
Spherical
130 Å
L1
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers