missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- HPLC Columns and Cartridges
- HPLC Columns
- Waters Corporation XBridge BEH Phenyl XP Column
Waters Corporation XBridge BEH Phenyl XP Column
Product Code. 15786072
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Quantity:
1/Ea.
3/Pk.
Unit Size:
Each
Pack of 3
22 product options available for selection
| Artikelnummer. | Length (Metric) | Diameter (Metric) | Quantity | unitSize |
|---|---|---|---|---|
| 15786072 | 50 mm | 4.6 mm | 1/Ea. | Each |
| 17113472 | 75 mm | 4.6 mm | 1/Ea. | Each |
| 17152502 | 30 mm | 2.1 mm | 3/Pk. | Pack of 3 |
| 17162502 | 50 mm | 2.1 mm | 3/Pk. | Pack of 3 |
| 17172502 | 75 mm | 2.1 mm | 3/Pk. | Pack of 3 |
| 17182502 | 100 mm | 2.1 mm | 3/Pk. | Pack of 3 |
| 17192502 | 30 mm | 3 mm | 3/Pk. | Pack of 3 |
| 17102512 | 50 mm | 3 mm | 3/Pk. | Pack of 3 |
| 17112512 | 75 mm | 3 mm | 3/Pk. | Pack of 3 |
| 17122512 | 100 mm | 3 mm | 3/Pk. | Pack of 3 |
| 17123462 | 30 mm | 2.1 mm | 1/Ea. | Each |
| 17133462 | 50 mm | 2.1 mm | 1/Ea. | Each |
| 17143462 | 75 mm | 2.1 mm | 1/Ea. | Each |
| 17153462 | 100 mm | 2.1 mm | 1/Ea. | Each |
| 17163462 | 30 mm | 3 mm | 1/Ea. | Each |
| 17173462 | 50 mm | 3 mm | 1/Ea. | Each |
| 17183462 | 75 mm | 3 mm | 1/Ea. | Each |
| 17193462 | 100 mm | 3 mm | 1/Ea. | Each |
| 17103472 | 30 mm | 4.6 mm | 1/Ea. | Each |
| 17123472 | 100 mm | 4.6 mm | 1/Ea. | Each |
| 17102602 | 150 mm | 2.1 mm | 3/Pk. | Pack of 3 |
| 17112602 | 150 mm | 3 mm | 3/Pk. | Pack of 3 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
Denne vare kan ikke returneres.
Se returpolicy
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
- This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
- Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Tekniske data
Tekniske data
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 50 mm |
| Max. Pressure | 10000 psi (690 Bar) |
| Particle Shape | Spherical |
| Particle Size | 2.5 μm |
| Pore Size | 130 Å |
| Quantity | 1/Ea. |
| USP Type | L11 |
| Endcapped | Yes |
| Type | eXtended Performance (XP) Column |
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?