missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge BEH HILIC XP Column

Product Code. 17162512
Encompass
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Quantity:
1/Ea.
3/Pk.
Unit Size:
Each
Pack of 3
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Quantity unitSize
17162512 100 mm 2.1 mm 3/Pk. Pack of 3
17143472 50 mm 2.1 mm 1/Ea. Each
17132512 30 mm 2.1 mm 3/Pk. Pack of 3
17142512 50 mm 2.1 mm 3/Pk. Pack of 3
17152512 75 mm 2.1 mm 3/Pk. Pack of 3
17172512 30 mm 3 mm 3/Pk. Pack of 3
17182512 50 mm 3 mm 3/Pk. Pack of 3
17192512 75 mm 3 mm 3/Pk. Pack of 3
17102522 100 mm 3 mm 3/Pk. Pack of 3
17133472 30 mm 2.1 mm 1/Ea. Each
17153472 75 mm 2.1 mm 1/Ea. Each
17163472 100 mm 2.1 mm 1/Ea. Each
17173472 30 mm 3 mm 1/Ea. Each
17183472 50 mm 3 mm 1/Ea. Each
17193472 75 mm 3 mm 1/Ea. Each
17103482 100 mm 3 mm 1/Ea. Each
17113482 30 mm 4.6 mm 1/Ea. Each
17123482 50 mm 4.6 mm 1/Ea. Each
17133482 75 mm 4.6 mm 1/Ea. Each
17143482 100 mm 4.6 mm 1/Ea. Each
17122602 150 mm 2.1 mm 3/Pk. Pack of 3
17132602 150 mm 3 mm 3/Pk. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Product Code. 17162512 Supplier Waters Corporation Supplier No. 176002581

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds.

  • These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 100 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 130 Å
Quantity 3/Pk.
USP Type L3
Endcapped No
Type eXtended Performance (XP) Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.