missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge BEH Amide XP Column

Product Code. 17193482
Encompass
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Quantity:
1/Ea.
3/Pk.
Unit Size:
Each
Pack of 3
Product Code. Length (Metric) Diameter (Metric) Quantity unitSize
17193482 75 mm 3 mm 1/Ea. Each
15441694 50 mm 3 mm 1/Ea. Each
15542701 100 mm 2.1 mm 1/Ea. Each
17142602 150 mm 2.1 mm 3/Pk. Pack of 3
17112522 30 mm 2.1 mm 3/Pk. Pack of 3
17122522 50 mm 2.1 mm 3/Pk. Pack of 3
17132522 75 mm 2.1 mm 3/Pk. Pack of 3
17142522 100 mm 2.1 mm 3/Pk. Pack of 3
17152522 30 mm 3 mm 3/Pk. Pack of 3
17162522 50 mm 3 mm 3/Pk. Pack of 3
17172522 75 mm 3 mm 3/Pk. Pack of 3
17182522 100 mm 3 mm 3/Pk. Pack of 3
17153482 30 mm 2.1 mm 1/Ea. Each
17163482 50 mm 2.1 mm 1/Ea. Each
17173482 75 mm 2.1 mm 1/Ea. Each
17183482 30 mm 3 mm 1/Ea. Each
17103492 100 mm 3 mm 1/Ea. Each
17113492 30 mm 4.6 mm 1/Ea. Each
17123492 50 mm 4.6 mm 1/Ea. Each
17133492 75 mm 4.6 mm 1/Ea. Each
17143492 100 mm 4.6 mm 1/Ea. Each
17152602 150 mm 3 mm 3/Pk. Pack of 3
22 options
This item is not returnable. View return policy

Product Code. 17193482

Brand: Waters Corporation 186006094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars.

  • XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.

Specifications

Diameter (Metric) 3 mm
Length (Metric) 75 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 130 Å
Quantity 1/Ea.
USP Type L68
Endcapped No
Type eXtended Performance (XP) Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.