missing translation for 'onlineSavingsMsg'
Learn More

XBP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18402717 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402717 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402717 Supplier Novus Biologicals Supplier No. NBP177681MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XBP1 Polyclonal antibody specifically detects XBP1 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen XBP1
Applications Flow Cytometry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human XBP1 protein (within residues 100-250). [Swiss-Prot P17861]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 7494
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.