missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XBP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£134.00
Specifications
| Antigen | XBP1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
XBP1 Polyclonal specifically detects XBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| XBP1 | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer, Lipid and Metabolism, Unfolded Protein Response | |
| PBS with 0.02% Sodium Azide | |
| TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
| XBP1 | |
| IgG | |
| Affinity Purified | |
| This antibody is raised to C-terminal of isoform XBP-1U (29kDa) and is not recognizing isoform XBP-1S (40kDa) |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P17861 | |
| 7494 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human XBP-1. The immunogen is located within the last 50 amino acids of XBP-1. Amino Acid Squence: LPAWRSSQRSTQKDPVPY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title