missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies XAV939
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- XAV939 is a potent, small molecule inhibitor of tankyrase (TNKS) 1 and 2 (IC50 = 11 and 4 nM, respectively) (Huang et al.) By inhibiting TNKS activity, XAV939 increases the protein levels of the axin-GSK3β complex and promotes the degradation of β-catenin in SW480 cells (Huang et al.), thereby inhibiting WNT pathway downstream actions
- Induces cardiomyogenesis in mesoderm progenitor cells derived from mouse embryonic stem cells (Wang et al.)
- In combination with the SMAD inhibitors LDN193189 and SB431542, promotes induction of forebrain fates in human pluripotent stem cell lines (Maroof et al.)
- Inhibits colony formation of APC-deficient, β-catenin-dependent DLD-1 colorectal cancer cells (Huang et al.)
Specifications
Specifications
| CAS | 284028-89-3 |
| Quantity | 1 mg |
| Molecular Formula | C14H11F3N2OS |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction