missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XAV 939, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | 1,5,7,8-Tetrahydro-2-[4-(trifluoromethyl)phenyl]-4H-thiopyrano[4,3-d]pyrimidin-4-one |
| CAS | 284028-89-3 |
| Molecular Formula | C14H11F3N2OS |
| Quantity | 25 mg |
| Synonym | XAV 939, NVP-XAV 939 |
| InChI Formula | InChI=1S/C14H11F3N2OS/c15-14(16,17)9-3-1-8(2-4-9)12-18-11-5-6-21-7-10(11)13(20)19-12/h1-4H,5-7H2,(H,18,19,20) |
| SMILES | Oc1nc(nc2CCSCc12)c3ccc(cc3)C(F)(F)F |
| IUPAC Name | 2-[4-(trifluoromethyl)phenyl]-7,8-dihydro-5H-thiopyrano[4,3-d]pyrimidin-4-ol |
| Molecular Weight (g/mol) | 312.31 |
| Formula Weight | 312.0544 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction