missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ XAP 044
GREENER_CHOICE

Product Code. 18796128
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18796128

Brand: Tocris Bioscience™ 5248/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective mGlu7 antagonist

XAP 044 is a potent and selective mGlu7 antagonist (IC50 = 88 nM); inhibits lateral amygdala long term potentiation (LTP) in brain slices from wild type mice. Displays no effects on the LTP of mGlu7 deficient mice. Exhibits novel activity by binding to the mGlu7 Venus flytrap domain (VFTD). Exhibits anti-anxiety effects in rodent models. Brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type XAP 044
Purity >98%
Molecular Weight (g/mol) 380.13
Inhibitors XAP 044

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.