missing translation for 'onlineSavingsMsg'
Learn More

Xanthydrol, TRC

Product Code. 30451246
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451246 2.5 g 2.5g
30449388 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451246 Supplier Toronto Research Chemicals Supplier No. TRCX7436502.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xanthydrol
CAS 90-46-0
Molecular Formula C13 H10 O2
Quantity 2.5 g
Synonym 9H-Xanthen-9-ol, 9-Hydroxyxanthene, 9-Xanthydrol, NSC 4038
InChI Formula InChI=1S/C13H10O2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8,13-14H
SMILES OC1c2ccccc2Oc3ccccc13
IUPAC Name 9H-xanthen-9-ol
Molecular Weight (g/mol) 198.22
Formula Weight 198.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.