missing translation for 'onlineSavingsMsg'
Learn More

Xanthurenic acid, MedChemExpress

Product Code. 16439376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439376 Supplier Medchem Express Supplier No. HYW014666
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Xanthurenic acid is a putative endogenous Group II metabotropic glutamate receptor agonist, on sensory transmission in the thalamus.

Xanthurenic acid is a putative endogenous Group II metabotropic glutamate receptor agonist, on sensory transmission in the thalamus.

Specifications

Chemical Name or Material Xanthurenic acid
CAS 59-00-7
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C10H7NO4
Quantity 100 mg
Solubility Information DMSO : 8.93 mg/mL (43.52 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=NC2=C(O)C=CC=C2C(O)=C1)O
Molecular Weight (g/mol) 205.17
Formula Weight 205.17
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.