missing translation for 'onlineSavingsMsg'
Learn More

Xanthosine Dihydrate, TRC

Product Code. 17904275
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904275 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17904275 Supplier Toronto Research Chemicals Supplier No. TRCX7421001G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5968-90-1
Molecular Formula C10 H12 N4 O6 . 2 H2 O
Quantity 1 g
Synonym 3,9-Dihydro-9-β-D-ribofuranosyl-1H-purine-2,6-dione Dihydrate, Xanthine Riboside Dihydrate;9-β-D-Ribofuranosylxanthine Dihydrate, NSC 18930 Dihydrate, Xanthine-9 β-D-Ribofuranoside Dihydrate
InChI Formula InChI=1S/C10H12N4O6.2H2O/c15-1-3-5(16)6(17)9(20-3)14-2-11-4-7(14)12-10(19)13-8(4)18;;/h2-3,5-6,9,15-17H,1H2,(H2,12,13,18,19);2*1H2/t3-,5-,6-,9-;;/m1../s1
SMILES O.O.OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3C(=O)NC(=O)Nc23
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;dihydrate
Molecular Weight (g/mol) 320.256
Formula Weight 320.0968
Chemical Name or Material Xanthosine Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.