missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xanthosine dihydrate, 99%

Product Code. 15636804 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15636804 1 g 1g
15646804 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15636804 Supplier Thermo Scientific Chemicals Supplier No. 207630010
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5968-90-1 | C10H12N4O6·2H2O | 320.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5968-90-1
Molecular Formula C10H12N4O6·2H2O
Molecular Weight (g/mol) 320.26
InChI Key AQQAMLPKPXTPOL-GWTDSMLYSA-N
Synonym xanthosine dihydrate, unii-1u62i480sp, 9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate, xanthosine, dihydrate, xanthosine dihydrate mi, xanthosine,dihydrate, 9-beta-d-ribofuranosyl xanthine dihydrate, 9-, a-d-ribofuranosylxanthine, xanthosine, dihydrate 8ci,9ci
PubChem CID 91886582
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 14,10065
Specific Rotation Condition −53.00 (22.00°C c=8, 0.3N NaOH)
Specific Rotation -53
Formula Weight 320.26
Percent Purity 99%
Chemical Name or Material Xanthosine dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.