missing translation for 'onlineSavingsMsg'
Learn More

Xanthone, Mikromol™

Product Code. 17980696
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980696 250 mg 250mg
17922796 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17980696 Supplier Mikromol Supplier No. MM1450.020250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Xanthone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xanthone
CAS 90-47-1
Analyte or Component Names Xanthone
Shipping Condition Room Temperature
Molecular Formula C13 H8 O2
Certifications/Compliance ISO 17034
Synonym Xanthen-9-one (8CI), 9-Oxoxanthene, 9-Xanthone, 9H-Xanthene, 9-oxo-, Benzophenone oxide, Diphenylene ketone oxide, Genicide, NSC 14978, Xanthenone, Xanthone, Dibenzo-gamma-pyrone
InChI Formula InChI=1S/C13H8O2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8H
SMILES O=C1c2ccccc2Oc3ccccc13
IUPAC Name xanthen-9-one
Molecular Weight (g/mol) 196.2
Quantity 250 mg
Formula Weight 196.0524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.