missing translation for 'onlineSavingsMsg'
Learn More

Xanthine, TRC

Product Code. 30451331
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451331 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451331 Supplier Toronto Research Chemicals Supplier No. TRCX49995010G
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xanthine
CAS 69-89-6
Purity Grade Notes HPLC
Molecular Formula C5 H4 N4 O2
Quantity 10 g
Synonym 1H-Purine-2,6-dione','3,9-dihydro-, 1H-Purine-2,6-dione','3,7-dihydro- (9CI), Xanthine (8CI), 1H,3H,7H-Xanthine, 1H,3H,9H-Xanthine, 1H-Purine-2,6-diol, 2,6-Dioxo-1,2,3,6-tetrahydropurine, 2,6-Dioxopurine, 3,9-Dihydro-1H-purine-2,6-dione, 3,9-Dihydropurine-2,6-dione, 9H-Purine-2,6(1H,3H)-dione, Isoxanthine, NSC 14664, Pseudoxanthine, Purine-2,6(1H,3H)-dione, Xan, Xanthic oxide, Xanthin
InChI Formula InChI=1S/C5H4N4O2/c10-4-2-3(7-1-6-2)8-5(11)9-4/h1H,(H3,6,7,8,9,10,11)
SMILES O=C1NC(=O)c2[nH]cnc2N1
IUPAC Name 3,7-dihydropurine-2,6-dione
Molecular Weight (g/mol) 152.11
Formula Weight 152.0334
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.