missing translation for 'onlineSavingsMsg'
Learn More

Xanthine Oxidase Antibody (8A9), Novus Biologicals™

Rabbit Monoclonal Antibody

£269.00 - £406.00

Specifications

Antigen Xanthine Oxidase
Clone 8A9
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30032440
View Documents
Novus Biologicals
NBP3-26250-50ul
50 μL
£269.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30032404
View Documents
Novus Biologicals
NBP3-26250-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Xanthine Oxidase Monoclonal antibody specifically detects Xanthine Oxidase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

Xanthine Oxidase
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
EC 1.17.1.4, EC 1.7.2.2, xanthene dehydrogenase, xanthine dehydrogenase, xanthine dehydrogenase/oxidase, xanthine oxidase, xanthine oxidoreductase, XDHA, XO, XOR
A synthesized peptide derived from Human Xanthine Oxidase [UniProt P47989]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
8A9
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Rabbit
Signal Transduction
PBS, pH 7.4, 150mM NaCl and 50% glycerol
7498
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.