missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthine monosodium salt monohydrate, ≥96% (dry basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Plastic bottle
Specifications
Physical Form | Crystalline Powder |
Quantity | 100 g |
Formula Weight | 192.11 |
Percent Purity | ≥96% |
Packaging | Plastic bottle |
Assay | Dry basis |
Chemical Name or Material | Xanthine Monosodium Salt Monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'