missing translation for 'onlineSavingsMsg'
Learn More

Xanthine, MedChemExpress

Product Code. 16403459
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16403459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16403459 Supplier Medchem Express Supplier No. HYW017389
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Xanthine, a plant alkaloid found in tea, coffee, and cocoa, is a mild stimulant of the central nervous system. Xanthine also acts as an intermediate product on the pathway of purine degradation.

Xanthine, a plant alkaloid found in tea, coffee, and cocoa, is a mild stimulant of the central nervous system. Xanthine also acts as an intermediate product on the pathway of purine degradation.

Specifications

Chemical Name or Material Xanthine
CAS 69-89-6
Color White
Purity Grade Notes Research
Molecular Formula C5H4N4O2
Quantity 1 g
Solubility Information 1M NaOH : 25 mg/mL (164.35 mM; ultrasonic and adjust pH to 12 with NaOH) ∣DMSO : 3.33 mg/mL (21.89 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N1)NC2=C(N=CN2)C1=O
Molecular Weight (g/mol) 152.11
Formula Weight 152.11
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.