missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthine, 99%, Thermo Scientific Chemicals
151.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.
Solubility
Soluble in water(0.067g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.
Solubility
Soluble in water(0.067g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | >300°C |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 8733 |
Merck Index | 14,10059 |
Solubility Information | Soluble in water(0.067g/L). |
Formula Weight | 152.11 |
Percent Purity | 99% |
Chemical Name or Material | Xanthine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'