missing translation for 'onlineSavingsMsg'
Learn More

Xanthene-9-carboxylic acid, 98%

Product Code. 11398395
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398395 25 g 25g
11388395 5 g 5g
11308405 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398395 Supplier Thermo Scientific Alfa Aesar Supplier No. L10384.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82-07-5 | C14H10O3 | 226.231 g/mol

Transesterification of the ethyl ester of commercially available xanthene-9-carboxylic acid yeildits 3- quinuclidinyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Transesterification of the ethyl ester of commercially available xanthene-9-carboxylic acid yeildits 3- quinuclidinyl ester.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-07-5
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00005059
InChI Key VSBFNCXKYIEYIS-UHFFFAOYSA-N
Synonym xanthene-9-carboxylic acid, xanthanoic acid, xanthenecarboxylic acid, 9-xanthenecarboxylic acid, unii-63a81f6a8v, xanthene-9-carboxylicacid, pubchem17697, maybridge1_006340, 9-xanthenylcarboxylic acid, chembl520
PubChem CID 65736
IUPAC Name 9H-xanthene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)C(=O)O

Specifications

Melting Point ∽217°C (decomposition)
Quantity 25 g
Beilstein 174414
Solubility Information Insoluble in water.
Formula Weight 226.23
Percent Purity 98%
Chemical Name or Material Xanthene-9-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.