missing translation for 'onlineSavingsMsg'
Learn More

Xanomeline oxalate, MedChemExpress

Product Code. p-200097812
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475668

Brand: Medchem Express HY13410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Xanomeline oxalate (LY246708 oxalate) is a potent and selective muscarinic receptor agonist (SMRA) and stimulates phosphoinositide hydrolysis in vivo. Xanomeline oxalate can be used for the research of Alzheimer’s disease.

Xanomeline oxalate (LY246708 oxalate) is a potent and selective muscarinic receptor agonist (SMRA) and stimulates phosphoinositide hydrolysis in vivo. Xanomeline oxalate can be used for the research of Alzheimer’s disease.

Specifications

Chemical Name or Material Xanomeline oxalate
Physical Form Solid
CAS 141064-23-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H25N3O5S
For Use With (Application) Neuroscience-Neurodegeneration
Synonym LY246708 oxalate
Solubility Information DMSO : 50 mg/mL (134.61 mM; Need ultrasonic)
SMILES CN1CCC=C(C2=NSN=C2OCCCCCC)C1.O=C(O)C(O)=O
Molecular Weight (g/mol) 371.45
Formula Weight 371.45
Percent Purity 96.26%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.