missing translation for 'onlineSavingsMsg'
Learn More

Xanomeline oxalate, MedChemExpress

Product Code. 16475668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475668 10 mg 10mg
16428367 5 mg 5mg
16421837 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475668 Supplier Medchem Express Supplier No. HY13410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Xanomeline oxalate (LY246708 oxalate) is a potent and selective muscarinic receptor agonist (SMRA) and stimulates phosphoinositide hydrolysis in vivo. Xanomeline oxalate can be used for the research of Alzheimer’s disease.

Xanomeline oxalate (LY246708 oxalate) is a potent and selective muscarinic receptor agonist (SMRA) and stimulates phosphoinositide hydrolysis in vivo. Xanomeline oxalate can be used for the research of Alzheimer’s disease.

Specifications

Chemical Name or Material Xanomeline oxalate
Physical Form Solid
CAS 141064-23-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H25N3O5S
For Use With (Application) Neuroscience-Neurodegeneration
Synonym LY246708 oxalate
Solubility Information DMSO : 50 mg/mL (134.61 mM; Need ultrasonic)
SMILES CN1CCC=C(C2=NSN=C2OCCCCCC)C1.O=C(O)C(O)=O
Molecular Weight (g/mol) 371.45
Formula Weight 371.45
Percent Purity 96.26%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.