missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XAGE2 Polyclonal Antibody

XAGE2 Polyclonal Antibody
Brand: Thermo Scientific PA562627
This item is not returnable.
View return policy
Description
XAGE2 Polyclonal Antibody for IHC
Specifications
| XAGE2 | |
| Polyclonal | |
| Unconjugated | |
| XAGE2 | |
| Cancer/testis antigen 12.2; cancer/testis antigen family 12, member 2; CT12.2; G antigen family D member 3; GAGED3; X antigen family member 2; X antigen family, member 2; X antigen family, member 2B; XAGE2; XAGE-2; XAGE2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9502 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GT9 | |
| XAGE2 | |
| Recombinant protein corresponding to Human XAGE2. Recombinant protein control fragment (Product #RP-104482). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction