missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XAGE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546413
This item is not returnable.
View return policy
Description
Peptide sequence: HERHTQTQNH TASPRSPVME SPKKKNQQLK VGILHLGSRQ KKIRIQLRSQ Sequence homology: Human: 100%.
This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in Ewing's sarcoma, alveolar rhabdomyosarcoma and normal testis. The protein encoded by this gene contains a nuclear localization signal and shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens. Alternative splicing of this gene, in addition to alternative transcription start sites, results in multiple transcript variants.
Specifications
| XAGE1 | |
| Polyclonal | |
| Unconjugated | |
| XAGE1B | |
| cancer/testis antigen 12.1; cancer/testis antigen family 12, member 1c; cancer/testis antigen family 12, member 1d; cancer/testis antigen family 12, member 1e; cancer/testis associated protein; CT12.1; CT12.1A; CT12.1C; CT12.1D; CT12.1E; CTP9; G antigen family D member 2; GAGED2; protein XAGE-1; X antigen family member 1; X antigen family member 1E; X antigen family, member 1C; X antigen family, member 1D; X antigen family, member 1E; XAGE1; XAGE-1; XAGE1A; XAGE1B; XAGE1C; XAGE1D; XAGE1E | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 653067, 653220 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HD64 | |
| XAGE1A, XAGE1B | |
| Synthetic peptide directed towards the middle region of human XAGE1B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction