missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XAF1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA141099

Product Code. 11518081

  • £441.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Suggested positive control: antigen standard for XAF1 (transient overexpression lysate), human liver tissue or protein. Immunogen sequence: NQTSTMEKDV RPKTRSIN & QLDLEKERY YKFKRFHF.

The inhibitors of apoptosis (IAP) proteins prevent cell death both in vitro and in vivo by virtue of inhibition of distinct caspases. The X-linked Inhibitor of Apoptosis, XIAP, is a key member of the IAP family of proteins. The caspase inhibiting activity of XIAP is negatively regulated by at least two XIAP-interacting proteins, XIAP-associated factor 1 (XAF1) and Smac/DIABLO. XAF1 binds to XIAP and antagonizes caspase inhibitory activities of XIAP. Expression of XAF1 triggers a redistribution of XIAP from the cytosol to the nucleus. XAF1 is ubiquitously expressed in normal tissues, but is present at low or undetectable levels in many different cancer cell lines.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

XAF1
Polyclonal
Unconjugated
XAF1
BIRC4 binding protein; BIRC4-binding protein; Birc4bp; F-box protein 39; Fbox39; HSXIAPAF1; novel TRAF-type zinc finger protein; XAF1; XIAP associated factor 1; XIAP associated factor-1; Xiapaf1; XIAP-associated factor 1
Rabbit
Protein G
RUO
327959, 54739
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q5NBU8, Q6GPH4
XAF1
Amino acids 201-218 and 301-317 of human XAF1.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.