missing translation for 'onlineSavingsMsg'
Learn More

XAF1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18405896 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405896 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405896 Supplier Novus Biologicals Supplier No. NBP176615AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XAF1 Polyclonal antibody specifically detects XAF1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen XAF1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias BIRC4-binding protein, BIRC4BPBIRC4 binding protein, HSXIAPAF1, XIAP associated factor 1, XIAPAF1XIAP-associated factor 1
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to amino acids at the C-terminus of human XAF-1. The immunogen is located within the last 50 amino acids of XAF-1. Amino Acid Squence: EKERYYKFKRFHF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 54739
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.