missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XAB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589444
This item is not returnable.
View return policy
Description
Immunogen sequence: PGQSGMDDMK LLEQRAEQLA AEAERDQPLR AQSKILFVRS DASREELAEL AQQVNPEEIQ LGEDEDEDEM DLEPNEVRLE QQSVPAAVFG SLKED; Positive Samples: U-87MG, A-549, LO2, HeLa; Cellular Location: Nucleus.
A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.
Specifications
| XAB2 | |
| Polyclonal | |
| Unconjugated | |
| XAB2 | |
| 0610041O14Rik; Adapter protein ATH-55; Ath55; AV025587; crn-related protein kim1; DKFZp762C1015; HCNP; HCNP protein; HCRN; KIAA1177; NTC90; PP3898; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; Xab2; XPA binding protein 2; XPA-binding protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56949, 67439 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.51 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9DCD2, Q9HCS7 | |
| XAB2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 761-855 of human XAB2 (NP_0645812). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction